C12H316CEB42 Chemcia Scientific, LLC

2-Iodo-5-methoxy-benzoic acid methyl ester

  • MDL number MFCD12923253
  • Cas Number : 857599-37-2
  • Molecular Weight : 292.07468

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3414

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3414
Availability details for
Quantity
  • Availability
  • Add to Cart