C12H316CEB4A Chemcia Scientific, LLC

4-Bromo-5-iodo-2-methyl-benzoic acid

  • MDL number MFCD28044132
  • Cas Number : 1022983-51-2
  • Molecular Weight : 340.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-iodo-2-methylbenzoic acid
Inchi Code 1S/C8H6BrIO2/c1-4-2-6(9)7(10)3-5(4)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key RVWSYPOXDJYFAQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3399

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3399
Availability details for
Quantity
  • Availability
  • Add to Cart