C12H316CEB4B Chemcia Scientific, LLC

2-Iodo-5-methyl-thiazole

  • MDL number MFCD20484739
  • Cas Number : 847547-16-4
  • Molecular Weight : 225.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-iodo-5-methyl-1,3-thiazole
Inchi Code 1S/C4H4INS/c1-3-2-6-4(5)7-3/h2H,1H3
Inchi Key HUSIZGWEEAVFKT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3397

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3397
Availability details for
Quantity
  • Availability
  • Add to Cart