C12H316CEB4C Chemcia Scientific, LLC

2-Isopropoxy-benzonitrile

  • MDL number MFCD02257394
  • Cas Number : 90921-35-0
  • Molecular Weight : 161.20527

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3394

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3394
Availability details for
Quantity
  • Availability
  • Add to Cart