C12H316CEB4D Chemcia Scientific, LLC

5-Chloro-2-hydroxy-4-methyl-benzoic acid methyl ester

  • MDL number MFCD06204186
  • Cas Number : 773134-18-2
  • Molecular Weight : 200.62328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3393

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3393
Availability details for
Quantity
  • Availability
  • Add to Cart