C12H316CEB51 Chemcia Scientific, LLC

5-Chloro-2-chloromethyl-3H-quinazolin-4-one

  • MDL number MFCD18089670
  • Molecular Weight : 229.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-(chloromethyl)-4(3H)-quinazolinone
Inchi Code 1S/C9H6Cl2N2O/c10-4-7-12-6-3-1-2-5(11)8(6)9(14)13-7/h1-4,12H,(H,13,14)
Inchi Key KATRZPFAMQFCJR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3424

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3424
Availability details for
Quantity
  • Availability
  • Add to Cart