C12H316CEB54 Chemcia Scientific, LLC

6-Bromo-2-chloromethyl-3H-quinazolin-4-one

  • MDL number MFCD06347511
  • Cas Number : 177167-05-4
  • Molecular Weight : 273.51797

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3427

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3427
Availability details for
Quantity
  • Availability
  • Add to Cart