C12H316CEB59 Chemcia Scientific, LLC

(3-Chloro-2-methyl-phenyl)-methyl-amine

  • MDL number MFCD10687534
  • Cas Number : 41456-52-4
  • Molecular Weight : 155.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-N,2-dimethylaniline
Inchi Code 1S/C8H10ClN/c1-6-7(9)4-3-5-8(6)10-2/h3-5,10H,1-2H3
Inchi Key XAEPSTFCZJTYLE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3432

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3432
Availability details for
Quantity
  • Availability
  • Add to Cart