C12H316CEB5B Chemcia Scientific, LLC

3-Amino-6-bromo-benzofuran-2-carboxylic acid methyl ester

  • MDL number MFCD27923239
  • Cas Number : 1823887-63-3
  • Molecular Weight : 270.08416

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3434

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3434
Availability details for
Quantity
  • Availability
  • Add to Cart