C12H316CEB5F Chemcia Scientific, LLC

3-Oxo-2,6,7,8-tetrahydro-3H-pyrido[3,2-c]pyridazine-5-carboxylic acid tert-butyl ester

  • MDL number MFCD28044137
  • Cas Number : 1701807-80-8
  • Molecular Weight : 251.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-oxo-2,3,6,7-tetrahydropyrido[3,2-c]pyridazine-5(1H)-carboxylate
Inchi Code 1S/C12H17N3O3/c1-12(2,3)18-11(17)15-6-4-5-8-9(15)7-10(16)14-13-8/h5,7,13H,4,6H2,1-3H3,(H,14,16)
Inchi Key ZCIYGWWMVQSNER-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3416

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3416
Availability details for
Quantity
  • Availability
  • Add to Cart