C12H316CEB60 Chemcia Scientific, LLC

3-Hydroxy-7,8-dihydro-6H-pyrido[3,2-c]pyridazine-5-carboxylic acid tert-butyl ester

  • MDL number MFCD28012411
  • Cas Number : 1701807-80-8
  • Molecular Weight : 251.28759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3417

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3417
Availability details for
Quantity
  • Availability
  • Add to Cart