C12H316CEB64 Chemcia Scientific, LLC

2-Hydroxy-3-nitro-benzonitrile

  • MDL number MFCD00234257
  • Cas Number : 28177-79-9
  • Molecular Weight : 164.12153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3443

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3443
Availability details for
Quantity
  • Availability
  • Add to Cart