C12H316CEB6E Chemcia Scientific, LLC

4-Chloro-3-iodo-1H-indazole

  • MDL number MFCD07781581
  • Cas Number : 518990-33-5
  • Molecular Weight : 278.48073

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3453

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3453
Availability details for
Quantity
  • Availability
  • Add to Cart