C12H316CEB72 Chemcia Scientific, LLC

3-Bromo-4-chloro-2H-indazole

  • MDL number MFCD28044139
  • Cas Number : 885521-40-4
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-chloro-2H-indazole
Inchi Code 1S/C7H4BrClN2/c8-7-6-4(9)2-1-3-5(6)10-11-7/h1-3H,(H,10,11)
Inchi Key KQVVWXSIPZGIAN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3458

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3458
Availability details for
Quantity
  • Availability
  • Add to Cart