C12H316CEB78 Chemcia Scientific, LLC

6-Benzyl-5,6,7,8-tetrahydro-3H-pyrido[4,3-d]pyrimidin-4-one

  • MDL number MFCD09835497
  • Cas Number : 109229-22-3
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-benzyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-4(1H)-one
Inchi Code 1S/C14H15N3O/c18-14-12-9-17(7-6-13(12)15-10-16-14)8-11-4-2-1-3-5-11/h1-5,10H,6-9H2,(H,15,16,18)
Inchi Key PVNGDFHKRAIVTC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3464

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3464
Availability details for
Quantity
  • Availability
  • Add to Cart