C12H316CEB7F Chemcia Scientific, LLC

2-Amino-6-chloro-benzoic acid methyl ester

  • MDL number MFCD16036437
  • Cas Number : 41632-04-6
  • Molecular Weight : 185.61146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3472

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3472
Availability details for
Quantity
  • Availability
  • Add to Cart