C12H316CEB81 Chemcia Scientific, LLC

2-Amino-4-iodo-nicotinic acid methyl ester

  • MDL number MFCD28044141
  • Cas Number : 1822818-22-3
  • Molecular Weight : 278.05044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3474

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3474
Availability details for
Quantity
  • Availability
  • Add to Cart