C12H316CEB84 Chemcia Scientific, LLC

3-Nitro-1H-pyrrolo[3,2-b]pyridine

  • MDL number MFCD11846273
  • Cas Number : 23612-33-1
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-nitro-1H-pyrrolo[3,2-b]pyridine
Inchi Code 1S/C7H5N3O2/c11-10(12)6-4-9-5-2-1-3-8-7(5)6/h1-4,9H
Inchi Key FTRQCCAMRDXXGH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3488

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3488
Availability details for
Quantity
  • Availability
  • Add to Cart