C12H316CEB89 Chemcia Scientific, LLC

2-Cyclohexyl-5-methyl-2,4-dihydro-pyrazol-3-one

  • MDL number MFCD00460245
  • Cas Number : 36210-76-1
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyclohexyl-5-methyl-2,4-dihydro-3H-pyrazol-3-one
Inchi Code 1S/C10H16N2O/c1-8-7-10(13)12(11-8)9-5-3-2-4-6-9/h7,9,11H,2-6H2,1H3
Inchi Key WVORGWGNCAZAES-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3501

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3501
Availability details for
Quantity
  • Availability
  • Add to Cart