C12H316CEB8E Chemcia Scientific, LLC

6-Methoxy-quinoxaline-2,3-diol

  • MDL number MFCD01825248
  • Cas Number : 31910-18-6
  • Molecular Weight : 192.17571

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3506

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3506
Availability details for
Quantity
  • Availability
  • Add to Cart