C12H316CEB90 Chemcia Scientific, LLC

2-Chloro-4-methoxy-5-nitro-benzoic acid methyl ester

  • MDL number MFCD26396226
  • Cas Number : 90537-46-5
  • Molecular Weight : 245.62081

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3508

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3508
Availability details for
Quantity
  • Availability
  • Add to Cart