C12H316CEB91 Chemcia Scientific, LLC

3-Amino-4-chloro-benzofuran-2-carboxylic acid methyl ester

  • MDL number MFCD28044149
  • Cas Number : 1139838-91-7
  • Molecular Weight : 225.63316

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3509

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3509
Availability details for
Quantity
  • Availability
  • Add to Cart