C12H316CEB9B Chemcia Scientific, LLC

2-Chloro-5-methyl-pyridine 1-oxide

  • MDL number MFCD11977414
  • Cas Number : 20173-49-3
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-methylpyridine 1-oxide
Inchi Code 1S/C6H6ClNO/c1-5-2-3-6(7)8(9)4-5/h2-4H,1H3
Inchi Key NYFBABAQZJZFED-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3519

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3519
Availability details for
Quantity
  • Availability
  • Add to Cart