C12H316CEBA4 Chemcia Scientific, LLC

1-Methyl-5-trifluoromethyl-1H-indazol-3-ylamine

  • MDL number MFCD07777192
  • Cas Number : 5685-69-8
  • Molecular Weight : 215.17941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3528

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3528
Availability details for
Quantity
  • Availability
  • Add to Cart