C12H316CEBB3 Chemcia Scientific, LLC

1-(2,2-Dimethoxy-ethyl)-2-isocyano-benzene

  • MDL number MFCD28044152
  • Cas Number : 592479-02-2
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,2-dimethoxyethyl)-2-isocyanobenzene
Inchi Code 1S/C11H13NO2/c1-12-10-7-5-4-6-9(10)8-11(13-2)14-3/h4-7,11H,8H2,2-3H3
Inchi Key XSTAOBURMHGXDG-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form Green Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3595

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3595
Availability details for
Quantity
  • Availability
  • Add to Cart