C12H316CEBB8 Chemcia Scientific, LLC

3-Iodo-2H-indazole-6-carboxylic acid methyl ester

  • MDL number MFCD23134632
  • Cas Number : 885518-82-1
  • Molecular Weight : 302.07274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3601

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3601
Availability details for
Quantity
  • Availability
  • Add to Cart