C12H316CEBB9 Chemcia Scientific, LLC

2-Amino-4-nitro-benzoic acid methyl ester

  • MDL number MFCD00089419
  • Cas Number : 3558-19-8
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3602

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3602
Availability details for
Quantity
  • Availability
  • Add to Cart