C12H316CEBBC Chemcia Scientific, LLC

2-Bromo-5-nitro-1H-indole-3-carbaldehyde

  • MDL number MFCD22570490
  • Cas Number : 1246471-79-3
  • Molecular Weight : 269.0558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3559

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3559
Availability details for
Quantity
  • Availability
  • Add to Cart