C12H316CEBC0 Chemcia Scientific, LLC

8-Bromo-5-fluoro-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid ethyl ester

  • MDL number MFCD22572834
  • Cas Number : 1000815-27-9
  • Molecular Weight : 314.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-bromo-5-fluoro-4-hydroxy-3-quinolinecarboxylate
Inchi Code 1S/C12H9BrFNO3/c1-2-18-12(17)6-5-15-10-7(13)3-4-8(14)9(10)11(6)16/h3-5H,2H2,1H3,(H,15,16)
Inchi Key FCMIOTBNVHXSJA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3583

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3583
Availability details for
Quantity
  • Availability
  • Add to Cart