C12H316CEBC2 Chemcia Scientific, LLC

8-Bromo-5-fluoro-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid

  • MDL number MFCD28044160
  • Cas Number : 1065093-62-0
  • Molecular Weight : 286.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-5-fluoro-4-hydroxy-3-quinolinecarboxylic acid
Inchi Code 1S/C10H5BrFNO3/c11-5-1-2-6(12)7-8(5)13-3-4(9(7)14)10(15)16/h1-3H,(H,13,14)(H,15,16)
Inchi Key PYLPYUFSELJLEK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3585

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3585
Availability details for
Quantity
  • Availability
  • Add to Cart