C12H316CEBCE Chemcia Scientific, LLC

2-Methoxymethoxy-4-nitro-phenylamine

  • MDL number MFCD15528115
  • Cas Number : 945969-40-4
  • Molecular Weight : 198.1799

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3617

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3617
Availability details for
Quantity
  • Availability
  • Add to Cart