C12H316CEBCF Chemcia Scientific, LLC

3-Iodo-pyridine-2,5-diamine

  • MDL number MFCD11844805
  • Cas Number : 1125410-09-4
  • Molecular Weight : 235.02807

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3618

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3618
Availability details for
Quantity
  • Availability
  • Add to Cart