C12H316CEBD0 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-2H-indazole-3-carboxylic acid ethyl ester

  • MDL number MFCD28044165
  • Cas Number : 174198-29-9
  • Molecular Weight : 194.23528

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3620

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3620
Availability details for
Quantity
  • Availability
  • Add to Cart