C12H316CEBD3 Chemcia Scientific, LLC

2-Amino-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid tert-butyl ester

  • MDL number MFCD02287893
  • Cas Number : 92932-02-0
  • Molecular Weight : 253.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate
Inchi Code 1S/C13H19NO2S/c1-13(2,3)16-12(15)10-8-6-4-5-7-9(8)17-11(10)14/h4-7,14H2,1-3H3
Inchi Key LBJBSHGWJTXGDG-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3623

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3623
Availability details for
Quantity
  • Availability
  • Add to Cart