C12H316CEBD5 Chemcia Scientific, LLC

3-(4-Fluoro-phenyl)-2,6-dihydro-4H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester

  • MDL number MFCD28044167
  • Cas Number : 1823314-67-5
  • Molecular Weight : 303.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(4-fluorophenyl)-2,6-dihydropyrrolo[3,4-c]pyrazole-5(4H)-carboxylate
Inchi Code 1S/C16H18FN3O2/c1-16(2,3)22-15(21)20-8-12-13(9-20)18-19-14(12)10-4-6-11(17)7-5-10/h4-7H,8-9H2,1-3H3,(H,18,19)
Inchi Key FEGGXUDUXKPQLC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3625

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3625
Availability details for
Quantity
  • Availability
  • Add to Cart