C12H316CEBD6 Chemcia Scientific, LLC

3-(4-Fluoro-phenyl)-4,6-dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester

  • MDL number MFCD28044168
  • Cas Number : 1823314-67-5
  • Molecular Weight : 303.33916

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3626

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3626
Availability details for
Quantity
  • Availability
  • Add to Cart