C12H316CEBD9 Chemcia Scientific, LLC

2-Amino-thiazolo[5,4-b]pyridin-5-ol

  • MDL number MFCD15526556
  • Cas Number : 31784-73-3
  • Molecular Weight : 167.19025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3629

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3629
Availability details for
Quantity
  • Availability
  • Add to Cart