C12H316CEBDB Chemcia Scientific, LLC

5-Methoxy-1H-thiazolo[5,4-b]pyridin-2-one

  • MDL number MFCD28044169
  • Cas Number : 1822818-32-5
  • Molecular Weight : 182.20207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3631

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3631
Availability details for
Quantity
  • Availability
  • Add to Cart