C12H316CEBDD Chemcia Scientific, LLC

2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyridine-3,6-dicarboxylic acid di-tert-butyl ester

  • MDL number MFCD28044171
  • Cas Number : 474843-69-1
  • Molecular Weight : 354.47177

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3633

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3633
Availability details for
Quantity
  • Availability
  • Add to Cart