C12H316CEBE0 Chemcia Scientific, LLC

2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid ethyl ester

  • MDL number MFCD26593859
  • Cas Number : 1297547-66-0
  • Molecular Weight : 196.20759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3636

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3636
Availability details for
Quantity
  • Availability
  • Add to Cart