C12H316CEBE1 Chemcia Scientific, LLC

2-Amino-5-bromo-4-nitro-benzoic acid methyl ester

  • MDL number MFCD28044172
  • Cas Number : 174566-52-0
  • Molecular Weight : 275.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-5-bromo-4-nitrobenzoate
Inchi Code 1S/C8H7BrN2O4/c1-15-8(12)4-2-5(9)7(11(13)14)3-6(4)10/h2-3H,10H2,1H3
Inchi Key OZQIKUAEJMHDBR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3637

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3637
Availability details for
Quantity
  • Availability
  • Add to Cart