C12H316CEBE3 Chemcia Scientific, LLC

1-(3-Dimethylamino-phenyl)-ethanone

  • MDL number MFCD00059343
  • Cas Number : 18992-80-8
  • Molecular Weight : 163.22121

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3639

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3639
Availability details for
Quantity
  • Availability
  • Add to Cart