C12H316CEBE4 Chemcia Scientific, LLC

5-Bromo-2-methylamino-benzonitrile

  • MDL number MFCD24231190
  • Cas Number : 106874-91-3
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-(methylamino)benzonitrile
Inchi Code 1S/C8H7BrN2/c1-11-8-3-2-7(9)4-6(8)5-10/h2-4,11H,1H3
Inchi Key SNCBXJLGIWYANH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3640

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3640
Availability details for
Quantity
  • Availability
  • Add to Cart