C12H316CEBE7 Chemcia Scientific, LLC

2-Chloro-6-iodo-benzoic acid methyl ester

  • MDL number MFCD18398956
  • Cas Number : 1261559-47-0
  • Molecular Weight : 296.49322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3644

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3644
Availability details for
Quantity
  • Availability
  • Add to Cart