C12H316CEBE9 Chemcia Scientific, LLC

(3-Fluoro-2-nitro-phenyl)-hydrazine

  • MDL number MFCD12149447
  • Cas Number : 435281-10-0
  • Molecular Weight : 171.13202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3646

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3646
Availability details for
Quantity
  • Availability
  • Add to Cart