C12H316CEBEF Chemcia Scientific, LLC

N-[2-(2,2-Dimethoxy-ethyl)-phenyl]-formamide

  • MDL number MFCD24564632
  • Cas Number : 592478-96-1
  • Molecular Weight : 209.2471

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3652

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3652
Availability details for
Quantity
  • Availability
  • Add to Cart