C12H316CEBF6 Chemcia Scientific, LLC

3-Nitro-2H-indazole

  • MDL number MFCD28044174
  • Cas Number : 863637-28-9
  • Molecular Weight : 163.1368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3746

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3746
Availability details for
Quantity
  • Availability
  • Add to Cart