C12H316CEC04 Chemcia Scientific, LLC

4-Bromo-3-thiocyanato-benzoic acid ethyl ester

  • MDL number MFCD28044177
  • Cas Number : 1823516-34-2
  • Molecular Weight : 286.14876

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3749

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3749
Availability details for
Quantity
  • Availability
  • Add to Cart