C12H316CEC05 Chemcia Scientific, LLC

4-Thiocyanato-2-trifluoromethoxy-phenylamine

  • MDL number MFCD11855816
  • Cas Number : 1133115-28-2
  • Molecular Weight : 234.20105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3750

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3750
Availability details for
Quantity
  • Availability
  • Add to Cart