C12H316CEC07 Chemcia Scientific, LLC

4-Bromo-5-methyl-1H-pyrazol-3-ylamine

  • MDL number MFCD03412169
  • Cas Number : 1780-72-9
  • Molecular Weight : 176.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-methyl-1H-pyrazol-3-amine
Inchi Code 1S/C4H6BrN3/c1-2-3(5)4(6)8-7-2/h1H3,(H3,6,7,8)
Inchi Key WZMBDMWFVPKYCF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3752

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3752
Availability details for
Quantity
  • Availability
  • Add to Cart